siRNA Details
Gene Symbol: Fam104a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034958.2 | NP_001030130.2 | 3 | 481 | ||
XM_006247695.2 | XP_006247757.1 | 4 | 561 | |||
XM_006247696.2 | XP_006247758.1 | 4 | 620 | |||
XM_006247697.2 | XP_006247759.1 | 4 | 459 | |||
GenBank mRNA | BC087161.1 | - | 3 | 323 | ||
BC105876.1 | - | 3 | 352 | 7129874 |

More Information
Other Species Targeted

