siRNA Details
Gene Symbol: LOC619574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034959.1 | NP_001030131.1 | 6 | 630 | ||
XM_006247911.2 | XP_006247973.1 | 6 | 676 | |||
GenBank mRNA | BC079126.1 | - | 6 | 630 | 7127052 |

More Information
Other Species Targeted

