siRNA Details
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034995.1 | NP_001030167.1 | 5 | 830 | ||
XM_006254636.2 | XP_006254698.1 | 5 | 924 | |||
XM_006254637.2 | XP_006254699.1 | 5 | 1238 | |||
XM_006254639.2 | XP_006254701.1 | 6 | 1035 | |||
XM_006254640.2 | XP_006254702.1 | 4 | 774 | |||
XM_006254641.2 | XP_006254703.1 | 3 | 440 | |||
XM_008772069.1 | XP_008770291.1 | 5 | 790 | |||
GenBank mRNA | BC103648.1 | - | 5 | 830 | 7315158 | |
BC166497.1 | - | 2 | 301 |
