siRNA Details
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034995.1 | NP_001030167.1 | 8 | 1600 | ||
XM_006254636.2 | XP_006254698.1 | 8 | 1694 | |||
XM_006254637.2 | XP_006254699.1 | 8 | 2008 | |||
XM_006254639.2 | XP_006254701.1 | 9 | 1805 | |||
XM_006254640.2 | XP_006254702.1 | 7 | 1544 | |||
XM_006254641.2 | XP_006254703.1 | 6 | 1210 | |||
XM_008772069.1 | XP_008770291.1 | 8 | 1560 | |||
GenBank mRNA | BC103648.1 | - | 8 | 1600 | 7315158 | |
BC166497.1 | - | 5 | 1071 | |||
FQ212790.1 | - | 2 | 194 |
