siRNA Details
Gene Symbol: Popdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037369.1 | NP_001032446.1 | 5 | 961 | ||
XM_006256599.2 | XP_006256661.1 | 4 | 1201 | |||
GenBank mRNA | BC105761.1 | - | 5 | 961 | 5623862 | |
FQ218039.1 | - | 5 | 864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
