siRNA Details
Gene Symbol: Defb51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037547.1 | NP_001032636.1 | 3 | 292 | ||
GenBank mRNA | AY621373.1 | - | 3 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
