siRNA Details
Gene Symbol: Defb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037503.1 | NP_001032592.1 | 2 | 85 | ||
GenBank mRNA | AY621345.1 | - | 2 | 85 |

More Information
Other Species Targeted

Gene Ontology Categories:
