siRNA Details
Gene Symbol: Defb24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037508.1 | NP_001032597.1 | 1, 2 | 113 | ||
GenBank mRNA | AY600148.1 | - | 1, 2 | 86 | ||
AY621355.1 | - | 1, 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
