siRNA Details
Gene Symbol: Defb21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037512.1 | NP_001032601.1 | 1, 2 | 65 | ||
GenBank mRNA | AY600147.1 | - | 1, 2 | 100 | ||
AY621352.1 | - | 1, 2 | 65 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
