siRNA Details
Gene Symbol: Defb25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037516.1 | NP_001032605.1 | 2 | 112 | ||
GenBank mRNA | AY621356.1 | - | 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
