siRNA Details
Gene Symbol: Defb33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037523.1 | NP_001032612.1 | 1, 2 | 112 | ||
XM_006253375.2 | XP_006253437.1 | 2, 3 | 236 | |||
GenBank mRNA | AY621362.1 | - | 1, 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
