siRNA Details
Gene Symbol: Rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037657.1 | NP_001032746.1 | 1761 | |||
XM_008762382.1 | XP_008760604.1 | 3 | 1821 | |||
GenBank mRNA | AY226099.1 | - | 1761 |
Gene Symbol: LOC100910990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235453.2 | XP_006235515.1 | 3 | 1789 | ||
XM_008762412.1 | XP_008760634.1 | 3 | 1821 | |||
XM_008762414.1 | XP_008760636.1 | 2 | 3784 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029397, NM_170598, AF345334, BC052473, AK129208, and others.
Gene Ontology Categories:
