siRNA Details
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100804.1 | NP_001094274.1 | 8 | 456 | ||
GenBank mRNA | AY262357.1 | - | 8 | 445 | ||
BC168196.1 | - | 456 | 7111460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
