siRNA Details
Gene Symbol: Fubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037653.1 | NP_001032742.1 | 15 | 1507 | ||
XM_006233507.2 | XP_006233569.1 | 16 | 1609 | |||
XM_006233511.2 | XP_006233573.1 | 16 | 1609 | |||
XM_006233512.2 | XP_006233574.1 | 16 | 1609 | |||
XM_006233513.2 | XP_006233575.1 | 15 | 1545 | |||
XM_008761553.1 | XP_008759775.1 | 16 | 1623 | |||
XM_008761554.1 | XP_008759776.1 | 16 | 1609 | |||
XM_008761555.1 | XP_008759777.1 | 15 | 1545 | |||
XM_008761556.1 | XP_008759778.1 | 15 | 1542 | |||
XM_008761557.1 | XP_008759779.1 | 15 | 1542 | |||
GenBank mRNA | BC107942.1 | - | 15 | 1507 | 7389449 |
