siRNA Details
Gene Symbol: Ndufa10l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182671.2 | NP_872612.1 | 1 | 192 | ||
GenBank mRNA | AY260163.1 | - | 1 | 192 |
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199495.2 | NP_955789.2 | 2 | 230 | ||
XM_008767385.1 | XP_008765607.1 | 2 | 242 | |||
GenBank mRNA | BC093375.1 | - | 2 | 189 | 7389276 | |
CB610197.1 | - | 2 | 230 | |||
FQ209525.1 | - | 2 | 232 |

More Information
Other Species Targeted

Gene Ontology Categories:
