siRNA Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109393.1 | NP_001102863.1 | 4 | 756 | ||
GenBank mRNA | FQ213131.1 | - | 4 | 493 | ||
FQ214060.1 | - | 4 | 464 |

More Information
Other Species Targeted

Gene Ontology Categories:
