siRNA Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109393.1 | NP_001102863.1 | 2 | 582 | ||
GenBank mRNA | FQ213131.1 | - | 2 | 319 | ||
FQ214060.1 | - | 2 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
