siRNA Details
Gene Symbol: LOC679038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100998.1 | NP_001094468.1 | 1 | 241 | ||
GenBank mRNA | BC105915.1 | - | 1 | 241 | 7384892 |
Gene Symbol: LOC678880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752079.3 | XP_003752127.1 | 1 | 315 |

More Information
Other Species Targeted

