siRNA Details
Gene Symbol: MYOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135940.1 | NP_001129412.1 | 8 | 877 | ||
NM_001300911.1 | NP_001287840.1 | 9 | 1008 | |||
NM_006790.2 | NP_006781.1 | 8 | 1359 | |||
XM_017010060.1 | XP_016865549.1 | 9 | 1577 | |||
XM_017010061.1 | XP_016865550.1 | 7 | 1010 | |||
XM_017010062.1 | XP_016865551.1 | 8 | 1446 | |||
GenBank mRNA | AF133820.2 | - | 8 | 1344 | ||
AF144477.1 | - | 8 | 1330 | |||
AK300076.1 | - | 9 | 988 | |||
AK300088.1 | - | 8 | 875 | |||
AK307620.1 | - | 9 | 1222 | |||
AK312733.1 | - | 8 | 1339 | |||
BC005376.1 | - | 8 | 1339 | 3934230 | ||
KU178476.1 | - | 7 | 1050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
