siRNA Details
Gene Symbol: Efcab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749810.3 | XP_003749858.1 | 2 | 1393 | ||
XM_008763265.1 | XP_008761487.1 | 2 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
