siRNA Details
Gene Symbol: Cox7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022503.2 | NP_071948.1 | 2 | 155 | ||
GenBank mRNA | BC166880.1 | - | 2 | 123 | 6888806 | |
BG668415.1 | - | 2 | 162 | |||
FQ209801.1 | - | 2 | 150 | |||
FQ209847.1 | - | 2 | 122 | |||
FQ210551.1 | - | 2 | 122 | |||
FQ211276.1 | - | 2 | 123 | |||
FQ214317.1 | - | 2 | 131 | |||
FQ217361.1 | - | 2 | 122 | |||
FQ221839.1 | - | 2 | 127 | |||
FQ223968.1 | - | 2 | 146 | |||
FQ228537.1 | - | 2 | 194 | |||
FQ229029.1 | - | 2 | 124 | |||
FQ229035.1 | - | 2 | 156 | |||
FQ230547.1 | - | 2 | 183 | |||
X54080.1 | - | 2 | 129 |
Gene Symbol: Cox7a2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751367.3 | XP_003751415.1 | 1 | 106 |
