siRNA Details
Gene Symbol: LOC679038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100998.1 | NP_001094468.1 | 1 | 721 | ||
GenBank mRNA | BC105915.1 | - | 1 | 721 | 7384892 |
Gene Symbol: LOC678880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752079.3 | XP_003752127.1 | 1 | 794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
