siRNA Details
Gene Symbol: Dmrtc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055549.5 | XP_001055549.2 | 6 | 793 | ||
XM_006257140.2 | XP_006257202.1 | 7 | 838 | |||
XM_008773329.1 | XP_008771551.1 | 6 | 3399 |
