siRNA Details
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127602.1 | NP_001121074.1 | 682 | |||
GenBank mRNA | BC166437.1 | - | 682 | 7383238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127602.1 | NP_001121074.1 | 682 | |||
GenBank mRNA | BC166437.1 | - | 682 | 7383238 |