siRNA Details
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127602.1 | NP_001121074.1 | 334 | |||
GenBank mRNA | BC166437.1 | - | 334 | 7383238 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001114179, NM_001082412, CX240158.
Gene Ontology Categories:
