siRNA Details
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127602.1 | NP_001121074.1 | 1001 | |||
GenBank mRNA | BC166437.1 | - | 1001 | 7383238 |

More Information
Other Species Targeted

Gene Ontology Categories:
