siRNA Details
Gene Symbol: LOC679149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100999.1 | NP_001094469.1 | 1107 | |||
GenBank mRNA | BC097486.1 | - | 1107 | 7459735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
