siRNA Details
Gene Symbol: LOC680288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001056467.3 | XP_001056467.1 | 1 | 124 |
Gene Symbol: LOC684509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749381.2 | XP_003749429.1 | 1 | 153 |

More Information
Other Species Targeted

