siRNA Details
Gene Symbol: Syce3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135253.1 | NP_001128725.1 | 2 | 433 | ||
XM_006242244.2 | XP_006242306.1 | 3 | 708 | |||
XM_006242246.2 | XP_006242308.1 | 3 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
