siRNA Details
Gene Symbol: Tmem169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109574.1 | NP_001103044.1 | 3 | 534 | ||
XM_006245284.2 | XP_006245346.1 | 2 | 445 | |||
XM_006245285.2 | XP_006245347.1 | 2 | 441 | |||
XM_006245286.2 | XP_006245348.1 | 3 | 467 | |||
XM_006245287.2 | XP_006245349.1 | 2 | 555 |
