siRNA Details
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101000.1 | NP_001094470.1 | 327 | |||
GenBank mRNA | BC105879.1 | - | 327 | 7123955 |

More Information
Other Species Targeted

Gene Ontology Categories:
