siRNA Details
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107401.1 | NP_001100871.1 | 1 | 418 | ||
XM_006254453.2 | XP_006254515.1 | 1 | 408 |

More Information
Other Species Targeted

