siRNA Details
Gene Symbol: Fam168b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271134.1 | NP_001258063.1 | 3 | 287 | ||
XM_008766974.1 | XP_008765196.1 | 2 | 883 | |||
XM_008766975.1 | XP_008765197.1 | 2 | 879 | |||
XM_008766976.1 | XP_008765198.1 | 2 | 881 | |||
XM_008766977.1 | XP_008765199.1 | 2 | 877 | |||
XM_008766978.1 | XP_008765200.1 | 3 | 331 |

More Information
Other Species Targeted

