siRNA Details
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109477.2 | NP_001102947.2 | 4, 5 | 264 | ||
XM_006240352.2 | XP_006240414.1 | 5, 6 | 473 | |||
GenBank mRNA | BC161941.1 | - | 303 | |||
EV765393.1 | - | 5, 6 | 303 | |||
FQ211665.1 | - | 268 |

More Information
Other Species Targeted

