siRNA Details
Gene Symbol: LOC680663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127377.1 | NP_001120849.1 | 10, 11 | 2007 | ||
XM_008773439.1 | XP_008771661.1 | 10, 11 | 2092 | |||
GenBank mRNA | BC166523.1 | - | 10, 11 | 2007 | 7134576 |
