siRNA Details
Gene Symbol: Sepsecs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128287.2 | NP_001121759.1 | 11 | 1349 | ||
GenBank mRNA | BC166584.1 | - | 11 | 1349 | 7443474 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172490, AK049338, AK033654, BC062133, AK153792.
Gene Ontology Categories:
