siRNA Details
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134702.1 | NP_001128174.1 | 6 | 607 | ||
XM_006251529.2 | XP_006251591.1 | 6 | 1855 | |||
GenBank mRNA | BC161984.1 | - | 6 | 607 | 9030600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
