siRNA Details
Gene Symbol: Ccdc176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134631.1 | NP_001128103.1 | 4 | 501 | ||
XM_006240348.2 | XP_006240410.1 | 4 | 499 | |||
XM_008764829.1 | XP_008763051.1 | 4 | 499 | |||
XM_008764830.1 | XP_008763052.1 | 4 | 372 |

More Information
Other Species Targeted

