siRNA Details
Gene Symbol: LOC100910957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254370.1 | XP_006254432.1 | 1 | 515 |
Gene Symbol: LOC679565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237717.2 | XP_006237779.1 | 1 | 655 |
