siRNA Details
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 628 | ||
NM_001270562.1 | NP_001257491.1 | 4 | 526 | |||
NM_001270563.1 | NP_001257492.1 | 4 | 486 | |||
NM_022593.4 | NP_072115.1 | 4 | 522 | |||
XM_006237724.2 | XP_006237786.1 | 4 | 523 | |||
GenBank mRNA | BC060566.1 | - | 4 | 497 | 6918038 | |
CK355540.1 | - | 4 | 522 | |||
FM093546.1 | - | 4 | 488 | |||
FM101371.1 | - | 4 | 476 | |||
FM110992.1 | - | 4 | 492 | |||
FQ213660.1 | - | 4 | 536 | |||
FQ214805.1 | - | 4 | 628 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026456, AK039746, AK011757, BC009104, AK011958, and others.
Gene Ontology Categories:
