siRNA Details
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014225.1 | NP_001014247.1 | 4, 5 | 747 | ||
XM_006245832.2 | XP_006245894.1 | 4, 5 | 510 | |||
XM_008767512.1 | XP_008765734.1 | 4, 5 | 443 | |||
XM_008767513.1 | XP_008765735.1 | 4, 5 | 805 | |||
XM_008767516.1 | XP_008765738.1 | 3, 4 | 349 | |||
XM_008767517.1 | XP_008765739.1 | 2, 3 | 212 | |||
XM_008767518.1 | XP_008765740.1 | 2, 3 | 269 | |||
GenBank mRNA | BC079022.1 | - | 4, 5 | 747 | 7113868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
