siRNA Details
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271214.1 | NP_001258143.1 | 11 | 1746 | ||
XM_008771657.1 | XP_008769879.1 | 11 | 1751 | |||
XM_008771658.1 | XP_008769880.1 | 12 | 2065 | |||
XM_008771659.1 | XP_008769881.1 | 12 | 2135 | |||
XM_008771660.1 | XP_008769882.1 | 10 | 1517 |
