siRNA Details
Gene Symbol: Shc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191065.1 | NP_001177994.1 | 2, 3 | 649 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_199022, AY464564.
Gene Ontology Categories:
