siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 870 | ||
NM_001278662.1 | NP_001265591.1 | 5 | 850 | |||
NM_001278663.1 | NP_001265592.1 | 3 | 647 | |||
NM_001278664.1 | NP_001265593.1 | 3 | 608 | |||
NM_001278677.1 | NP_001265606.1 | 6 | 942 | |||
NM_001278678.1 | NP_001265607.1 | 2 | 512 | |||
NM_203282.3 | NP_975011.3 | 4 | 735 | |||
XM_011528443.1 | XP_011526745.1 | 6 | 1003 | |||
XM_011528444.1 | XP_011526746.1 | 7 | 1023 | |||
XM_011528448.2 | XP_011526750.1 | 4 | 682 | |||
XM_017027514.1 | XP_016883003.1 | 6 | 933 | |||
XM_017027515.1 | XP_016883004.1 | 8 | 1360 | |||
XM_017027516.1 | XP_016883005.1 | 4 | 3407 | |||
XM_017027517.1 | XP_016883006.1 | 5 | 1236 | |||
XM_017027518.1 | XP_016883007.1 | 4 | 732 | |||
XM_017027519.1 | XP_016883008.1 | 5 | 1011 | |||
GenBank mRNA | AF054180.1 | - | 4 | 721 | ||
AK291286.1 | - | 4 | 702 | |||
AK303431.1 | - | 3 | 581 | |||
BC043147.1 | - | 4 | 707 | 5296940 | ||
BC139749.1 | - | 4 | 666 | 40115903 | ||
DA195951.1 | - | 3 | 466 | |||
DA230395.1 | - | 2 | 508 | |||
DA501852.1 | - | 1 | 264 |
