siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 963 | ||
NM_001278662.1 | NP_001265591.1 | 5 | 943 | |||
NM_001278663.1 | NP_001265592.1 | 3 | 740 | |||
NM_001278664.1 | NP_001265593.1 | 3 | 701 | |||
NM_001278677.1 | NP_001265606.1 | 6 | 1035 | |||
NM_001278678.1 | NP_001265607.1 | 2 | 605 | |||
NM_203282.3 | NP_975011.3 | 4 | 828 | |||
XM_011528443.1 | XP_011526745.1 | 6 | 1096 | |||
XM_011528444.1 | XP_011526746.1 | 7 | 1116 | |||
XM_011528448.2 | XP_011526750.1 | 4 | 775 | |||
XM_017027514.1 | XP_016883003.1 | 6 | 1026 | |||
XM_017027515.1 | XP_016883004.1 | 8 | 1453 | |||
XM_017027516.1 | XP_016883005.1 | 4 | 3500 | |||
XM_017027517.1 | XP_016883006.1 | 5 | 1329 | |||
XM_017027518.1 | XP_016883007.1 | 4 | 825 | |||
XM_017027519.1 | XP_016883008.1 | 5 | 1104 | |||
GenBank mRNA | AF054180.1 | - | 4 | 814 | ||
AK291286.1 | - | 4 | 795 | |||
AK303431.1 | - | 3 | 674 | |||
BC043147.1 | - | 4 | 800 | 5296940 | ||
BC139749.1 | - | 4 | 759 | 40115903 | ||
DA195951.1 | - | 3 | 559 | |||
DA230395.1 | - | 2 | 601 | |||
DA501852.1 | - | 1 | 357 |
