siRNA Details
Gene Symbol: Gpr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079710.1 | NP_001073178.1 | 2 | 527 | ||
GenBank mRNA | BC128782.1 | - | 2 | 527 | 7375738 |

More Information
Other Species Targeted

Gene Ontology Categories:
