siRNA Details
Gene Symbol: Tceal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_577026.6 | XP_577026.4 | 3 | 350 | ||
GenBank mRNA | FQ213674.1 | - | 3 | 327 | ||
FQ213749.1 | - | 3 | 288 |
Gene Symbol: Tceal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749245.3 | XP_003749293.1 | 3 | 362 | ||
GenBank mRNA | FQ213604.1 | - | 3 | 288 |

More Information
Other Species Targeted

