siRNA Details
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055430.5 | XP_001055430.4 | 3, 4 | 835 | ||
XM_006241414.2 | XP_006241476.1 | 4, 5 | 429 | |||
XM_008765427.1 | XP_008763649.1 | 3, 4 | 855 | |||
XM_008765428.1 | XP_008763650.1 | 4, 5 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
