siRNA Details
Gene Symbol: Mab21l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109391.1 | NP_001102861.1 | 1 | 1910 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011839, BC046628, AK053645, AF126427, AK144373, and others.
Gene Ontology Categories:
