siRNA Details
Gene Symbol: LOC680112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109392.1 | NP_001102862.1 | 2 | 335 | ||
XM_006236383.2 | XP_006236445.1 | 3 | 307 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC680112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109392.1 | NP_001102862.1 | 2 | 335 | ||
XM_006236383.2 | XP_006236445.1 | 3 | 307 |